Learn More
Microsolv Technology Corporation Vials, Headspace, Crimp Top, Glass. Clear. 9ml, 20mm Serum Finish, 19x50mm outer dimensions, Beveled Top and Rounded Bottom with a Tapered Neck. Perkin Elmer Compatible. 100/PK
Catalog No. 502269910
Encompass_Preferred
This item has a minimum order quantity of 10 packs per supplier requirements.
First hydrolytic, Class A, Borosilicate Glass. Use with aluminum or steel crimp caps for proper sealing. Compatible with Perkin Elmer, Tekmar and other Headspace autosamplers.
This item is not returnable. View return policy
Catalog No. 50-226-9910 Supplier Microsolv Technology Corporation Supplier No. 95025EPE09C
This item has a minimum order quantity of 10 packs per supplier requirements.
First hydrolytic, Class A, Borosilicate Glass. Use with aluminum or steel crimp caps for proper sealing. Compatible with Perkin Elmer, Tekmar and other Headspace autosamplers.