Learn More
Microsolv Technology Corporation Vials, Headspace, Crimp Top, Glass. Clear. 12ml, 20mm Serum Finish, 19x65mm outer dimensions, Beveled Top and Rounded Bottom with a Tapered Neck. Perkin Elmer Compatible. 100/PK.
Catalog No. 502269912
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass. Use with aluminum or steel crimp caps for proper sealing. Compatible with Perkin Elmer, Tekmar and other Headspace autosamplers.
This item is not returnable. View return policy
Catalog No. 50-226-9912 Supplier Microsolv Technology Corporation Supplier No. 95025EPE12C
First hydrolytic, Class A, Borosilicate Glass. Use with aluminum or steel crimp caps for proper sealing. Compatible with Perkin Elmer, Tekmar and other Headspace autosamplers.