Learn More
Microsolv Technology Corporation Vials, Headspace, Crimp Top, Glass. Clear, 27ml, 20mm Serum Finish, 30x60mm outer dimensions, Beveled Top and Flat Bottom with a Tapered Neck. 100/PK.
Catalog No. 502269908
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, Use with aluminum or steel crimp caps for proper sealing.
This item is not returnable. View return policy
Catalog No. 50-226-9908 Supplier Microsolv Technology Corporation Supplier No. 95025EAP27C
First hydrolytic, Class A, Borosilicate Glass, Use with aluminum or steel crimp caps for proper sealing.