Learn More
Microsolv Technology Corporation Vials, Headspace, Crimp Top, Glass. Clear, 20ml, 20mm Serum Finish, 22.5x75.5mm outer dimensions, Flat Bottom with a Long Neck. Agilent Compatible. 100/PK.
Catalog No. 502270276
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, Use with aluminum or steel crimp caps for proper sealing. Compatible with Agilent, Carlo Erba, Dani and other Headspace autosamplers.
This item is not returnable. View return policy
Catalog No. 50-227-0276 Supplier Microsolv Technology Corporation Supplier No. 95025AG20C
First hydrolytic, Class A, Borosilicate Glass, Use with aluminum or steel crimp caps for proper sealing. Compatible with Agilent, Carlo Erba, Dani and other Headspace autosamplers.