Learn More
Microsolv Technology Corporation Vials, Headspace, Crimp Top, Glass. Clear. 20ml, 20mm Serum Finish, 23x75.5mm outer dimensions, Beveled Top and Rounded Bottom with a Tapered Neck. Perkin Elmer Compatible. 100/PK.
Catalog No. 502270278
Encompass_Preferred
First hydrolytic, Class A, Type N51A pharmaceutical grade, Borosilicate Glass. Use with aluminum or steel crimp caps for proper sealing. Compatible with Perkin Elmer, Tekmar and other Headspace autosamplers.
This item is not returnable. View return policy
Catalog No. 50-227-0278 Supplier Microsolv Technology Corporation Supplier No. 95025PE20C
First hydrolytic, Class A, Type N51A pharmaceutical grade, Borosilicate Glass. Use with aluminum or steel crimp caps for proper sealing. Compatible with Perkin Elmer, Tekmar and other Headspace autosamplers.