Learn More
Microsolv Technology Corporation Vials, Headspace, Crimp Top, Glass. Clear, 20ml, 20mm Serum Finish, 23x75mm outer dimensions, Beveled Top and Flat Bottom with a Tapered Neck. Agilent Compatible. 100/PK.
Catalog No. 502270965
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, Use with aluminum or steel crimp caps for proper sealing.
This item is not returnable. View return policy
Catalog No. 50-227-0965 Supplier Microsolv Technology Corporation Supplier No. 95025AP20C
First hydrolytic, Class A, Borosilicate Glass, Use with aluminum or steel crimp caps for proper sealing.