Learn More
Microsolv Technology Corporation Septa, 20mm, Headspace, Molded Butyl Rubber, Dark Grey. For use in 20mm Crimp Caps, sold separately. 1000/CS.
Supplier:  Microsolv Technology Corporation 9502501
Encompass_Preferred
Shaped, Butyl Rubber Septa, Durometer of 55 Shore A, (hardness) and 3.0mm thick. Temperature resistant from -40⁰ to 120⁰C. Use in Aluminum or Steel Crimp Caps for Headspace autosamplers. Useful when PTFE is a contaminant or one of the analytes. Supplied in polyethylene bags. Can also be used as a stopper.
Catalog No. 50-227-1197