Learn More
Microsolv Technology Corporation Vials, Headspace, Screw Top, Glass. Amber, 20ml, 18mm precision threads, 22.5x75.5mm outer dimensions, Rounded Bottom. 1000/CS.
Catalog No. 502270269
Encompass_Preferred
First hydrolytic, Class A, Type N51A pharmaceutical grade, Borosilicate Glass, amberized and considered low actinic. Features 18mm precision threads for proper sealing. Compatible with CTC Combi Pal, Perkin Elmer, Agilent and other Headspace autosamplers. Not suitable for older versions of TurboMatrix 110 that were manufactured before 2006.
This item is not returnable. View return policy
Catalog No. 50-227-0269 Supplier Microsolv Technology Corporation Supplier No. 950200AVK
First hydrolytic, Class A, Type N51A pharmaceutical grade, Borosilicate Glass, amberized and considered low actinic. Features 18mm precision threads for proper sealing. Compatible with CTC Combi Pal, Perkin Elmer, Agilent and other Headspace autosamplers. Not suitable for older versions of TurboMatrix 110 that were manufactured before 2006.