Learn More
Microsolv Technology Corporation Vials, Headspace, Screw Top, Glass. Amber, 10ml, 18mm precision threads, 22.5x46mm outer dimensions, Rounded Bottom. CTC Compliant. 100/PK.
Catalog No. 502270956
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, amberized and considered low actinic. Features 18mm precision threads for proper sealing. Compatible with CTC, LEAP Combi Pal, Agilent and other Headspace autosamplers. Not suitable for older versions of TurboMatrix 110 that were manufactured before 2006. Lowest cost in class.
This item is not returnable. View return policy
Catalog No. 50-227-0956 Supplier Microsolv Technology Corporation Supplier No. 95010E0AV
First hydrolytic, Class A, Borosilicate Glass, amberized and considered low actinic. Features 18mm precision threads for proper sealing. Compatible with CTC, LEAP Combi Pal, Agilent and other Headspace autosamplers. Not suitable for older versions of TurboMatrix 110 that were manufactured before 2006. Lowest cost in class.