Learn More
Microsolv Technology Corporation Vials, Headspace, Screw Top, Glass. Amber, 10ml, 18mm precision threads, 22.5x46mm outer dimensions, Rounded Bottom. CTC Compliant. 100/PK.
Supplier:  Microsolv Technology Corporation 950100AV
Encompass_Preferred
First hydrolytic, Class A, Type N51A pharmaceutical grade, Borosilicate Glass, amberized and considered low actinic. Features 18mm precision threads for proper sealing. Compatible with CTC, LEAP Combi Pal, Agilent and other Headspace autosamplers. Not suitable for older versions of TurboMatrix 110 that were manufactured before 2006.
Catalog No. 50-227-0265