Learn More
Microsolv Technology Corporation Vials Hs 10Ml Screw Top Rd Clr
Catalog No. 5001451821
Encompass_Preferred
Vials, Headspace, Screw Top, Glass. Clear, 10ml, 18mm precision threads, 22.5x46mm outer dimensions, Rounded Bottom. CTC Compliant. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51821 Supplier Microsolv Technology Corporation Supplier No. 950100CV
Vials, Headspace, Screw Top, Glass. Clear, 10ml, 18mm precision threads, 22.5x46mm outer dimensions, Rounded Bottom. CTC Compliant. MicroSolv Brand. 100/PK.