Learn More
Microsolv Technology Corporation Vials Hs 20Ml Screw Top Rd Clr
Catalog No. 5001451829
Encompass_Preferred
Vials, Headspace, Screw Top, Glass. Clear, 20ml, 18mm precision threads, 22.5x75.5mm outer dimensions, Rounded Bottom. CTC Compliant. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51829 Supplier Microsolv Technology Corporation Supplier No. 950200CV
Vials, Headspace, Screw Top, Glass. Clear, 20ml, 18mm precision threads, 22.5x75.5mm outer dimensions, Rounded Bottom. CTC Compliant. MicroSolv Brand. 100/PK.