Learn More
Microsolv Technology Corporation Vials Max Rec 3.8 Trumpet Btm
Catalog No. 5001452394
Encompass_Preferred
Vials, Screw Top, Glass. Clear, 3.8ml, Max Recovery™, 13-425mm threads, 14.75x45mm outer dimensions for 13mm screw caps. For use as an autosampler vial or storage bottle. AQ Brand. 100/PK.
Catalog No. 50-014-52394 Supplier Microsolv Technology Corporation Supplier No. 9534S0CV
Vials, Screw Top, Glass. Clear, 3.8ml, Max Recovery™, 13-425mm threads, 14.75x45mm outer dimensions for 13mm screw caps. For use as an autosampler vial or storage bottle. AQ Brand. 100/PK.