Learn More
Microsolv Technology Corporation Vials Pp Screw 300Ul
Supplier:  Microsolv Technology Corporation 9532SMS
Encompass_Preferred
Vials, Screw Top, Plastic. Clear, 300ul MicroVial, LCMS Compatible, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. AQ Brand. 100/PK.
Catalog No. 50-014-52389