Learn More
Microsolv Technology Corporation Vials Pp Screw 700Ul
Catalog No. 5001452397
Encompass_Preferred
Vials, Screw Top, Plastic. 700ul, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52397 Supplier Microsolv Technology Corporation Supplier No. 9572SPP
Vials, Screw Top, Plastic. 700ul, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 100/PK.