Learn More
Microsolv Technology Corporation Vials Pp Snap 300Ul. Lcms
Catalog No. 5001452364
Encompass_Preferred
Vials, Snap Top, Plastic. Clear, 300ul MicroVial, LCMS Compatible. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. AQ Brand. 100/PK.
Catalog No. 50-014-52364 Supplier Microsolv Technology Corporation Supplier No. 9532CMS
Vials, Snap Top, Plastic. Clear, 300ul MicroVial, LCMS Compatible. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. AQ Brand. 100/PK.