Learn More
Microsolv Technology Corporation Vials Pp Snap 300Ulvials Pp S
Catalog No. 5001452353
Encompass_Preferred
Vials, Snap Top, Plastic. Clear, 300ul MicroVial. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52353 Supplier Microsolv Technology Corporation Supplier No. 9532C0PV
Vials, Snap Top, Plastic. Clear, 300ul MicroVial. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 100/PK.