Learn More
Chemglass Life Sciences Kit, Clear 2.0mL 12x32mm 9mm Thread Large Opening Vial and a PTFE/Silicone Septa Lined BONDED CLOSURE
SDP
Catalog No. NC0526847
Encompass_Preferred
Kit, Clear 2.0mL 12x32mm 9mm Thread Large Opening Vial and a PTFE/Silicone Septa Lined BONDED CLOSURE. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.
Catalog No. NC0526847 Supplier Chemglass Life Sciences Supplier No. CV51681232
Kit, Clear 2.0mL 12x32mm 9mm Thread Large Opening Vial and a PTFE/Silicone Septa Lined BONDED CLOSURE. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.