Learn More
Microsolv Technology Corporation Vials, Push Top, Storage, Glass, Clear, 5ml. Economical, with ND18 tops. For use with Econo-Storage Push Top Caps. 100/PK.
Catalog No. 502271373
Encompass_Preferred
These Vials are made with 3rd Hydrolytic Glass and quickly open and reseal due to the "Snap" design. These 5ml Vials have a total fill volume of 9.1ml with 40x20mm outer dimensions. Not made for Crimp Caps. Caps for these Vials, sold separately. These vials are not intended for Analytical Testing and should only be used for Storage. Low cost for budget minded laboratories.
This item is not returnable. View return policy
Catalog No. 50-227-1373 Supplier Microsolv Technology Corporation Supplier No. 97010CV05
These Vials are made with 3rd Hydrolytic Glass and quickly open and reseal due to the "Snap" design. These 5ml Vials have a total fill volume of 9.1ml with 40x20mm outer dimensions. Not made for Crimp Caps. Caps for these Vials, sold separately. These vials are not intended for Analytical Testing and should only be used for Storage. Low cost for budget minded laboratories.