Learn More
Microsolv Technology Corporation Vials Rsa Clear Write On W/Fus
Catalog No. 5001452350
Encompass_Preferred
Vials, Snap Top, Glass. Clear / Clear Insert, 300ul with Write-On Patch and fill lines, Fused Insert™. An 11mm snap ring and 12x32mm outer dimensions. For use as an autosampler vial. RSA Brand. 100/PK.
Catalog No. 50-014-52350 Supplier Microsolv Technology Corporation Supplier No. 9532C0CVRS
Vials, Snap Top, Glass. Clear / Clear Insert, 300ul with Write-On Patch and fill lines, Fused Insert™. An 11mm snap ring and 12x32mm outer dimensions. For use as an autosampler vial. RSA Brand. 100/PK.