Learn More
Microsolv Technology Corporation Vials Rsa Clear Write On W/Fus
Catalog No. 5001452371
Encompass_Preferred
Vials, Screw Top, Glass. Clear / Clear Insert, 300ul with Write-On Patch and fill lines, Fused Insert™, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. RSA Brand. 100/PK.
Catalog No. 50-014-52371 Supplier Microsolv Technology Corporation Supplier No. 9532S0CVRS
Vials, Screw Top, Glass. Clear / Clear Insert, 300ul with Write-On Patch and fill lines, Fused Insert™, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. RSA Brand. 100/PK.