Learn More
Microsolv Technology Corporation Vials Scre Clear Silanizedvia
Catalog No. 5001452094
Encompass_Preferred
Vials, Screw Top, Glass. Clear, 2ml, Deactivated, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52094 Supplier Microsolv Technology Corporation Supplier No. 9502S0DCV
Vials, Screw Top, Glass. Clear, 2ml, Deactivated, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 100/PK.