Learn More
Neta Scientific 2 ML Volume, Screw Top Closure, Glass, Amber, 12 Mm Cap Size, 12 Mm Width, 32 Mm Height,
SDP
Catalog No. 50929364
Encompass_Preferred
This item is manufactured by Agilent
Agilent Vial, screw top, amber, write-on spot, certified, 2 mL, 100/pk. Vial size: 12 x 32 mm (12 mm cap) - 2 ml Screw Top Vials. (Caps not included)
Catalog No. 50-929-364 Supplier Neta Scientific Supplier No. 51820716
This item is manufactured by Agilent
Agilent Vial, screw top, amber, write-on spot, certified, 2 mL, 100/pk. Vial size: 12 x 32 mm (12 mm cap) - 2 ml Screw Top Vials. (Caps not included)