Learn More
Microsolv Technology Corporation Vials Screw Clear 1.5Ml
Catalog No. 5001452092
Encompass_Preferred
Vials, Screw Top, Glass. Clear, 2ml, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52092 Supplier Microsolv Technology Corporation Supplier No. 9502S0CV
Vials, Screw Top, Glass. Clear, 2ml, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. MicroSolv Brand. 100/PK.