Learn More
Microsolv Technology Corporation Vials Screw Clear
Catalog No. 5001452158
Encompass_Preferred
Vials, Screw Top, Glass. Clear, 2ml, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. BASIK Brand. 100/PK.
Catalog No. 50-014-52158 Supplier Microsolv Technology Corporation Supplier No. 9502SE0CV
Vials, Screw Top, Glass. Clear, 2ml, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. BASIK Brand. 100/PK.