Learn More
Microsolv Technology Corporation Vials Screw Clear Write On
Catalog No. 5001452193
Encompass_Preferred
Vials, Screw Top, Glass. Clear, 2ml, with Write-On Patch and fill lines, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52193 Supplier Microsolv Technology Corporation Supplier No. 9502SWCV
Vials, Screw Top, Glass. Clear, 2ml, with Write-On Patch and fill lines, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. MicroSolv Brand. 100/PK.