Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Amber / Clear Insert, 300ul Fused Insert™, 10-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502270034
Encompass_Preferred
Wide mouth vials and inserts are first hydrolytic, Class A, Borosilicate Glass, 300ul total fill volume, inserts fused to the bottom of the vial. Amber vials are often used for light sensitive compounds and are considered low actinic. Our most economical Fused-Insert Vials with 10mm screw threads. For budget minded laboratories.
This item is not returnable. View return policy
Catalog No. 50-227-0034 Supplier Microsolv Technology Corporation Supplier No. 9533SE0AV
Wide mouth vials and inserts are first hydrolytic, Class A, Borosilicate Glass, 300ul total fill volume, inserts fused to the bottom of the vial. Amber vials are often used for light sensitive compounds and are considered low actinic. Our most economical Fused-Insert Vials with 10mm screw threads. For budget minded laboratories.