Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Amber / Clear Insert, 300ul Fused Insert, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502271100
Encompass_Preferred
Vials and inserts are first hydrolytic, Class A, Borosilicate Glass, with a wide bottom, 300ul, inserts fused to the bottom of the vial. The wide bottom helps prevent bubble formation when filling quickly or with automation. Our most economical Fused-Insert Vials. For budget minded laboratories.
This item is not returnable. View return policy
Catalog No. 50-227-1100 Supplier Microsolv Technology Corporation Supplier No. 9532SE0AV
Vials and inserts are first hydrolytic, Class A, Borosilicate Glass, with a wide bottom, 300ul, inserts fused to the bottom of the vial. The wide bottom helps prevent bubble formation when filling quickly or with automation. Our most economical Fused-Insert Vials. For budget minded laboratories.