Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Clear / Clear Insert, 300ul with Write-On Patch and fill lines, Wide Tip Fused Insert™, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502270575
Encompass_Preferred
Vials and Wide Tip Inserts are made with first hydrolytic, Class A, Type-33 Clear, Borosilicate Glass. The insert has a 300ul total fill volume and a 4ul residual volume for many autosamplers and needles. The Write-On Patch and fill lines are ceramic and will resist smearing or scratching even when working with solvents. The Wide Tip Insert bottom depth, heights and alignment are designed, and QC controlled for maximum recovery and needle safety. The wide bottom helps prevent bubble formation when filling quickly or using with automation. This product is manufactured in an ISO 9001:2015 Certified facility. Favorite choice of many clinical testing laboratories
This item is not returnable. View return policy
Catalog No. 50-227-0575 Supplier Microsolv Technology Corporation Supplier No. 9532SWCVWT
Vials and Wide Tip Inserts are made with first hydrolytic, Class A, Type-33 Clear, Borosilicate Glass. The insert has a 300ul total fill volume and a 4ul residual volume for many autosamplers and needles. The Write-On Patch and fill lines are ceramic and will resist smearing or scratching even when working with solvents. The Wide Tip Insert bottom depth, heights and alignment are designed, and QC controlled for maximum recovery and needle safety. The wide bottom helps prevent bubble formation when filling quickly or using with automation. This product is manufactured in an ISO 9001:2015 Certified facility. Favorite choice of many clinical testing laboratories