Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Clear / Clear Insert, 300ul with Write-On Patch and fill lines, Fused Insert™, Deactivated, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502270574
Encompass_Preferred
Vial and Inserts are made with first hydrolytic, Class A, Type 33 Clear, Silanized, Borosilicate Glass. The Write-On Patch and fill lines are ceramic and will resist smearing or scratching even when working with solvents.Insert bottom depth, heights and alignment are designed, and QC controlled for maximum recovery and needle safety. This product is manufactured in an ISO 9001:2015 Certified facility. The AQ™ Brand Fused Inserts sit slightly above the top lip of the vial. This unique feature allows for a near perfect seal with the cap's septa. These inserts are not attached to the bottom of the vial and do not suffer from cracking at this fused point and prevents leaking which can be observed in competitive brands. Consistency from vial to vial is one feature that sets these vials apart from other similar silanized products. Good choice for low abundance analytes, some proteins and peptides. Compares to Q-Serts™ and other “Certified” Brands.
This item is not returnable. View return policy
Catalog No. 50-227-0574 Supplier Microsolv Technology Corporation Supplier No. 9532S0DCV
Vial and Inserts are made with first hydrolytic, Class A, Type 33 Clear, Silanized, Borosilicate Glass. The Write-On Patch and fill lines are ceramic and will resist smearing or scratching even when working with solvents.Insert bottom depth, heights and alignment are designed, and QC controlled for maximum recovery and needle safety. This product is manufactured in an ISO 9001:2015 Certified facility. The AQ™ Brand Fused Inserts sit slightly above the top lip of the vial. This unique feature allows for a near perfect seal with the cap's septa. These inserts are not attached to the bottom of the vial and do not suffer from cracking at this fused point and prevents leaking which can be observed in competitive brands. Consistency from vial to vial is one feature that sets these vials apart from other similar silanized products. Good choice for low abundance analytes, some proteins and peptides. Compares to Q-Serts™ and other “Certified” Brands.