Learn More
Microsolv Technology Corporation Vials, Screw Top 2Ml Amber Pp
Catalog No. NC1005012
Encompass_Preferred
Vials, Screw Top, Plastic. Amber, 2ml, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 100/PK.
Catalog No. NC1005012 Supplier Microsolv Technology Corporation Supplier No. 9502SPPAM
Vials, Screw Top, Plastic. Amber, 2ml, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 100/PK.