Learn More
Sigma Aldrich Fine Chemicals Biosciences Vials screw top clear glas
Catalog No. 502900004
Encompass
Vials clear glass (1.5mL) are suitable basically to inject samples from an autosampler. Autosampler needle pierces through the cap during injection and withdraws the required aliquots of sample from the vials.
Catalog No. 50-290-0004 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 854171
Vials clear glass (1.5mL) are suitable basically to inject samples from an autosampler. Autosampler needle pierces through the cap during injection and withdraws the required aliquots of sample from the vials.