Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Clear, 2ml, with Write-On Patch and fill lines, 8-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. 100/PK.
Catalog No. 5001452413
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass. Write-On Patch and fill lines are epoxy paint which will resist smearing during use. Features a narrow mouth which may be useful to reduce evaporation
Catalog No. 50-014-52413 Supplier Microsolv Technology Corporation Supplier No. 9582SWCV
First hydrolytic, Class A, Borosilicate Glass. Write-On Patch and fill lines are epoxy paint which will resist smearing during use. Features a narrow mouth which may be useful to reduce evaporation