Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Amber, 2ml, with Write-On Patch and fill lines, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502270039
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, amberized. Features a wide mouth for easy filling. Low cost for budget minded laboratories. Amber vials are often used for light sensitive compounds and considered low actinic. Write-On Patch and fill lines are epoxy paint for reduced smearing with solvents. Lowest cost in class for budget minded laboratories.
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Catalog No. 50-227-0039 Supplier Microsolv Technology Corporation Supplier No. 9502SEWAV
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
First hydrolytic, Class A, Borosilicate Glass, amberized. Features a wide mouth for easy filling. Low cost for budget minded laboratories. Amber vials are often used for light sensitive compounds and considered low actinic. Write-On Patch and fill lines are epoxy paint for reduced smearing with solvents. Lowest cost in class for budget minded laboratories.
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'