Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Amber, 2ml, with Write-On Patch and graduations, 10-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502271080
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, amberized. Features a wide mouth opening for easy filling and ND10 threads. Write-On Patch and fill lines are epoxy paint for reduced smearing. Amber vials are often used for light sensitive compounds and considered low actinic.
This item is not returnable. View return policy
Catalog No. 50-227-1080 Supplier Microsolv Technology Corporation Supplier No. 9503SEWAV
First hydrolytic, Class A, Borosilicate Glass, amberized. Features a wide mouth opening for easy filling and ND10 threads. Write-On Patch and fill lines are epoxy paint for reduced smearing. Amber vials are often used for light sensitive compounds and considered low actinic.