Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Amber, 2ml, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502271077
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, amberized. Features a wide mouth for easy filling. Lowest cost in class. Amber vials are often used for light sensitive compounds and considered low actinic.
This item is not returnable. View return policy
Catalog No. 50-227-1077 Supplier Microsolv Technology Corporation Supplier No. 9502SE0AV
First hydrolytic, Class A, Borosilicate Glass, amberized. Features a wide mouth for easy filling. Lowest cost in class. Amber vials are often used for light sensitive compounds and considered low actinic.