Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Amber, 4ml, with Write-On Patch and graduations, 13-425mm threads, 15x45mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502271082
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, amberized. Features ND13 threads. Write-On Patch is epoxy paint for reduced smearing. Amber vials are often used for light sensitive compounds and considered low actinic.
This item is not returnable. View return policy
Catalog No. 50-227-1082 Supplier Microsolv Technology Corporation Supplier No. 9504SEWAV
First hydrolytic, Class A, Borosilicate Glass, amberized. Features ND13 threads. Write-On Patch is epoxy paint for reduced smearing. Amber vials are often used for light sensitive compounds and considered low actinic.