Learn More
Microsolv Technology Corporation Vials, Screw Top, Storage, Glass, Amber, 12ml, with 15-425mm Screw Threads. For use as a Storage Vial. 100/PK.
Catalog No. 502271383
Encompass_Preferred
First hydrolytic, Class A, Type N51A, Pharmaceutical Grade, Borosilicate Glass, amberized. Amber vials are often used for light sensitive compounds and are considered low actinic. These Storage Vials have 66x18.5mm outer dimensions and a total fill volume of 12ml and can be used for long term storage with a proper cap. Use with Caps that have 15-425 or ND15 screw threads, sold separately.
This item is not returnable. View return policy
Catalog No. 50-227-1383 Supplier Microsolv Technology Corporation Supplier No. 97015AV12
First hydrolytic, Class A, Type N51A, Pharmaceutical Grade, Borosilicate Glass, amberized. Amber vials are often used for light sensitive compounds and are considered low actinic. These Storage Vials have 66x18.5mm outer dimensions and a total fill volume of 12ml and can be used for long term storage with a proper cap. Use with Caps that have 15-425 or ND15 screw threads, sold separately.