Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Amber, 2ml, with Write-On Patch and fill lines, Deactivated, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502271612
Encompass_Preferred
First hydrolytic, Class A, Deactivated / Silanized Borosilicate Glass, amberized. Features a wide mouth for easy filling. Low cost for budget minded laboratories. Write-On Patch and fill lines are epoxy paint for reduced smearing with solvents. Amber vials are often used for light sensitive compounds and considered low actinic.
This item is not returnable. View return policy
Catalog No. 50-227-1612 Supplier Microsolv Technology Corporation Supplier No. 9502SEWDAV
First hydrolytic, Class A, Deactivated / Silanized Borosilicate Glass, amberized. Features a wide mouth for easy filling. Low cost for budget minded laboratories. Write-On Patch and fill lines are epoxy paint for reduced smearing with solvents. Amber vials are often used for light sensitive compounds and considered low actinic.