Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Amber, 4ml, 13-425mm threads, 15x45mm outer dimensions. For use as an autosampler vial or storage bottle. 100/PK.
Catalog No. 502271374
Encompass_Preferred
First hydrolytic, Class A, Type N51A pharmaceutical grade, Borosilicate Glass, amberized. Features ND13 threads. Amber vials are often used for light sensitive compounds and considered low actinic. Use these vials with Waters WISP and older Shimadzu autosamplers.
This item is not returnable. View return policy
Catalog No. 50-227-1374 Supplier Microsolv Technology Corporation Supplier No. 9504S0AV
First hydrolytic, Class A, Type N51A pharmaceutical grade, Borosilicate Glass, amberized. Features ND13 threads. Amber vials are often used for light sensitive compounds and considered low actinic. Use these vials with Waters WISP and older Shimadzu autosamplers.