Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Amber, 2ml, with Write-On Patch and fill lines, 8-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. 100/PK.
Catalog No. 502270038
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, amberized. Write-On Patch and fill lines are epoxy paint which will resist smearing during use. Features a narrow mouth which may be useful to reduce evaporation and requires an 8mm screw cap. Amber vials are often used for light sensitive compounds and considered low actinic.
This item is not returnable. View return policy
Catalog No. 50-227-0038 Supplier Microsolv Technology Corporation Supplier No. 9582SWAV
First hydrolytic, Class A, Borosilicate Glass, amberized. Write-On Patch and fill lines are epoxy paint which will resist smearing during use. Features a narrow mouth which may be useful to reduce evaporation and requires an 8mm screw cap. Amber vials are often used for light sensitive compounds and considered low actinic.