Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Amber, 2ml, with Write-On Patch and fill lines, Deactivated, 10-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502270515
Encompass_Preferred
First hydrolytic, Class A, Type N51A, Deactivated / Silanized pharmaceutical grade, Borosilicate Glass, amberized. Features a wide mouth opening for easy filling and ND10 threads. Write-On Patch and fill lines are epoxy paint for reduced smearing. Amber vials are often used for light sensitive compounds and are considered low actinic.
This item is not returnable. View return policy
Catalog No. 50-227-0515 Supplier Microsolv Technology Corporation Supplier No. 9503SWDAV
First hydrolytic, Class A, Type N51A, Deactivated / Silanized pharmaceutical grade, Borosilicate Glass, amberized. Features a wide mouth opening for easy filling and ND10 threads. Write-On Patch and fill lines are epoxy paint for reduced smearing. Amber vials are often used for light sensitive compounds and are considered low actinic.