Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Amber, 1.8ml, Max Recovery™, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502270572
Encompass_Preferred
First hydrolytic, Class A, Type N51A, Reduced Surface Activity, Borosilicate Glass, amberized. Amber glass is often used for light sensitive compounds and is considered low actinic. This market leading 1.8ml vial has the low residual volume of 8ul due to the center-draining effects of smooth walls (no ledges) and a conical bottom, which allows needles with bottom entry ports to recover the maximum amount during use. Bottom depths are designed and QC controlled for the most critical autosampler to prevent needles hitting the bottom of the vials. This product is manufactured in an ISO 9001:2015 Certified facility. This is a very popular 1.8ml, center draining autosampler vial because of the consistency of the glass chemistry from batch to batch, vial to vial. Reduced surface activity is tested for each lot and certified. Excellent for low abundance analytes, pH sensitive analytes and for LCMS.
This item is not returnable. View return policy
Catalog No. 50-227-0572 Supplier Microsolv Technology Corporation Supplier No. 9512S0AVRS
First hydrolytic, Class A, Type N51A, Reduced Surface Activity, Borosilicate Glass, amberized. Amber glass is often used for light sensitive compounds and is considered low actinic. This market leading 1.8ml vial has the low residual volume of 8ul due to the center-draining effects of smooth walls (no ledges) and a conical bottom, which allows needles with bottom entry ports to recover the maximum amount during use. Bottom depths are designed and QC controlled for the most critical autosampler to prevent needles hitting the bottom of the vials. This product is manufactured in an ISO 9001:2015 Certified facility. This is a very popular 1.8ml, center draining autosampler vial because of the consistency of the glass chemistry from batch to batch, vial to vial. Reduced surface activity is tested for each lot and certified. Excellent for low abundance analytes, pH sensitive analytes and for LCMS.