Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Amber, 2ml, with Write-On Patch and fill lines, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. 100/PK.
Catalog No. 502270308
Encompass_Preferred
First hydrolytic, Class A, Type N51A, Borosilicate Glass, amberized. Write-On Patch and fill lines are ceramic and will resist smearing or scratching when working with solvents. Compares to HPLC “Certified” vials for performance but lower priced and a great value. Bottom depths and heights are designed, and QC controlled for maximum recovery and needle safety. Amber vials are often used for light sensitive compounds and considered low actinic.
This item is not returnable. View return policy
Catalog No. 50-227-0308 Supplier Microsolv Technology Corporation Supplier No. 9509SWAVAQ
First hydrolytic, Class A, Type N51A, Borosilicate Glass, amberized. Write-On Patch and fill lines are ceramic and will resist smearing or scratching when working with solvents. Compares to HPLC “Certified” vials for performance but lower priced and a great value. Bottom depths and heights are designed, and QC controlled for maximum recovery and needle safety. Amber vials are often used for light sensitive compounds and considered low actinic.