Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Amber, 2ml, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. 1,000/CS.
Catalog No. 502271076
Encompass_Preferred
First hydrolytic, Class A, pharmaceutical grade, Borosilicate Glass, amberized. Features a wide mouth opening for easy filling and ND9 threads. Lowest cost in class. Case quantity is the most economical value.
This item is not returnable. View return policy
Catalog No. 50-227-1076 Supplier Microsolv Technology Corporation Supplier No. 9502S0AVK
First hydrolytic, Class A, pharmaceutical grade, Borosilicate Glass, amberized. Features a wide mouth opening for easy filling and ND9 threads. Lowest cost in class. Case quantity is the most economical value.