Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Amber, 2ml, Deactivated, 8-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. 100/PK.
Catalog No. 502270037
Encompass_Preferred
First hydrolytic, Class A, Deactivated / Silanized, Borosilicate Glass. Features a narrow mouth which may be useful to reduce evaporation and requires an 8mm screw cap. Amber vials are often chosen for light sensitive compounds and is considered low actinic. Good choice for biological compounds such as proteins and peptides due to the hydrophobic nature of the vial.
This item is not returnable. View return policy
Catalog No. 50-227-0037 Supplier Microsolv Technology Corporation Supplier No. 9582S0DAV
First hydrolytic, Class A, Deactivated / Silanized, Borosilicate Glass. Features a narrow mouth which may be useful to reduce evaporation and requires an 8mm screw cap. Amber vials are often chosen for light sensitive compounds and is considered low actinic. Good choice for biological compounds such as proteins and peptides due to the hydrophobic nature of the vial.