Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Clear, 2ml, 10-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502271083
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass. Features a wide mouth for easy filling and ND10 threads.
This item is not returnable. View return policy
Catalog No. 50-227-1083 Supplier Microsolv Technology Corporation Supplier No. 9503SE0CV
First hydrolytic, Class A, Borosilicate Glass. Features a wide mouth for easy filling and ND10 threads.