Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Clear, 4ml, 13-425mm threads, 15x45mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502271085
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass. Features ND13 threads. Low cost for budget minded laboratories.
This item is not returnable. View return policy
Catalog No. 50-227-1085 Supplier Microsolv Technology Corporation Supplier No. 9504SE0CV
First hydrolytic, Class A, Borosilicate Glass. Features ND13 threads. Low cost for budget minded laboratories.