Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Clear, 2ml, with Write-On Patch and fill lines, Deactivated, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Supplier:  Microsolv Technology Corporation 9502SWDCV
Encompass_Preferred
First hydrolytic, Class A, Type N51A pharmaceutical grade, Borosilicate Deactivated / Silanized Glass. Write-On Patch and fill lines are epoxy paint for reduced smearing. Features a wide mouth opening for easy filling and ND9 threads.
Catalog No. 50-227-0517