Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Clear, 1.2ml, MRQ™, Deactivated, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Supplier:  Microsolv Technology Corporation 9512S0CVTD
Encompass_Preferred
First hydrolytic, Class A, Type-33 Deactivated / Silanized, Borosilicate Glass. The MRQ Vials have a fill volume of 1.2ml and a very low residual volume of 2ul due to the center-draining effects of smooth walls (no ledges) and a conical bottom with a well that is slightly larger than autosampler vial needles, with side entry ports to recover the maximum amount during use. Bottom depths of the wells are designed and QC controlled for the most critical autosampler to prevent needles hitting the bottom of the vials. This product is manufactured in an ISO 9001:2015 Certified facility.
Catalog No. 50-227-0577